siRNA Details
| Gene Symbol: TRABD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080824.2 | NP_001074293.1 | 3 | 934 | ||
| NM_001277053.1 | NP_001263982.1 | 4 | 1081 | |||
| NM_001307978.1 | NP_001294907.1 | 4 | 1081 | |||
| XM_011532504.1 | XP_011530806.1 | 4 | 881 | |||
| GenBank mRNA | AK296608.1 | - | 4 | 918 | ||
| AK309615.1 | - | 4 | 920 | |||
| BC049209.1 | - | 3 | 621 | |||
| BC051789.1 | - | 3 | 935 | |||
| BC073808.1 | - | 3 | 358 | |||
| JQ653415.1 | - | 4 | 875 | |||

TRABD2A
Genomic Map