siRNA Details
| Gene Symbol: FAM172BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036433.2 | 2 | 483 | |||
| GenBank mRNA | AK127584.1 | - | 2 | 484 | ||
| EG328077.1 | - | 1 | 216 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM172BP| Gene Symbol: FAM172BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036433.2 | 2 | 483 | |||
| GenBank mRNA | AK127584.1 | - | 2 | 484 | ||
| EG328077.1 | - | 1 | 216 | |||