siRNA Details
| Gene Symbol: ARL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206919.1 | NP_996802.1 | 3 | 514 | ||
| XM_005265719.3 | XP_005265776.1 | 3 | 628 | |||
| XM_005265720.3 | XP_005265777.1 | 3 | 696 | |||
| XM_006714036.3 | XP_006714099.1 | 3 | 388 | |||
| GenBank mRNA | AY439003.1 | - | 3 | 514 | ||

ARL9
Genomic Map