siRNA Details
| Gene Symbol: LOC105374344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134676.1 | 3 | 432 | |||
| GenBank mRNA | AA813499.1 | - | 1 | 61 | ||
| BX116031.1 | - | 3 | 432 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374344| Gene Symbol: LOC105374344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134676.1 | 3 | 432 | |||
| GenBank mRNA | AA813499.1 | - | 1 | 61 | ||
| BX116031.1 | - | 3 | 432 | |||