siRNA Details
| Gene Symbol: TENM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122679.1 | NP_001116151.1 | 1 | 211 | ||
| XM_005265950.2 | XP_005266007.1 | 3 | 858 | |||
| XM_006714897.2 | XP_006714960.1 | 3 | 858 | |||
| XM_011534604.2 | XP_011532906.1 | 3 | 858 | |||
| XM_017009660.1 | XP_016865149.1 | 3 | 858 | |||
| XM_017009661.1 | XP_016865150.1 | 3 | 858 | |||
| XM_017009662.1 | XP_016865151.1 | 3 | 858 | |||
| GenBank mRNA | HE578282.1 | - | 1 | 211 | ||
| HE578283.1 | - | 1 | 211 | |||

TENM2
Genomic Map