siRNA Details
| Gene Symbol: IRAK1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010844.3 | NP_001010844.1 | 1 | 72 | ||
| XM_006715336.3 | XP_006715399.1 | 1 | 113 | |||
| XM_011535447.1 | XP_011533749.1 | 1 | 112 | |||
| XM_011535448.2 | XP_011533750.1 | 1 | 130 | |||
| XM_017010265.1 | XP_016865754.1 | 1 | 130 | |||
| GenBank mRNA | BC112253.1 | - | 1 | 61 | 8327721 | |
| BC112255.1 | - | 1 | 61 | 8327723 | ||
| CR990080.1 | - | 1 | 72 | |||

IRAK1BP1
Genomic Map