siRNA Details
| Gene Symbol: PM20D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010853.2 | NP_001010853.1 | 6, 7 | 1242 | ||
| XM_005248661.2 | XP_005248718.1 | 7, 8 | 1241 | |||
| XM_005248662.4 | XP_005248719.1 | 7, 8 | 992 | |||
| XM_011535481.2 | XP_011533783.1 | 6, 7 | 1474 | |||
| XM_017010290.1 | XP_016865779.1 | 6, 7 | 1064 | |||
| XM_017010291.1 | XP_016865780.1 | 7, 8 | 884 | |||
| GenBank mRNA | AK127529.1 | - | 4, 5 | 537 | ||
| AK302464.1 | - | 6, 7 | 1202 | |||
| BC035036.1 | - | 6, 7 | 1245 | 5262128 | ||
| BC045583.1 | - | 6, 7 | 993 | |||
| BX640865.1 | - | 1, 2 | 37 | |||

PM20D2
Genomic Map