siRNA Details
| Gene Symbol: CCDC26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130917.1 | 4 | 704 | |||
| NR_130918.1 | 3 | 481 | ||||
| NR_130919.1 | 4 | 635 | ||||
| NR_130920.1 | 4 | 652 | ||||
| GenBank mRNA | BC026098.1 | - | 3 | 482 | ||
| BC070152.1 | - | 4 | 704 | |||
| BG393734.1 | - | 4 | 609 | |||
| BG532915.1 | - | 3 | 481 | |||
| BU186641.1 | - | 4 | 626 | |||

CCDC26
Genomic Map