siRNA Details
| Gene Symbol: HGSNAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152419.2 | NP_689632.2 | 7, 8 | 801 | ||
| XM_005273409.1 | XP_005273466.1 | 7, 8 | 801 | |||
| XM_005273410.1 | XP_005273467.1 | 7, 8 | 801 | |||
| XM_005273411.1 | XP_005273468.1 | 7, 8 | 801 | |||
| XM_005273412.3 | XP_005273469.1 | 7, 8 | 801 | |||
| XM_017013088.1 | XP_016868577.1 | 7, 8 | 801 | |||
| XM_017013089.1 | XP_016868578.1 | 7, 8 | 705 | |||
| GenBank mRNA | AK304441.1 | - | 7, 8 | 753 | ||
| BC042037.1 | - | 9, 10 | 864 | |||
| CR749838.1 | - | 3, 4 | 225 | |||

HGSNAT
Genomic Map