siRNA Details
| Gene Symbol: C9orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128618.1 | NP_001122090.1 | 5 | 535 | ||
| XM_011518225.2 | XP_011516527.1 | 4 | 379 | |||
| GenBank mRNA | BC036255.1 | - | 5 | 535 | ||
| BC044620.1 | - | 5 | 513 | |||
| BC053956.1 | - | 5 | 513 | |||
| BC130404.1 | - | 4 | 434 | 40146435 | ||
| BC144273.1 | - | 4 | 433 | 9052793 | ||

C9orf57
Genomic Map