siRNA Details
| Gene Symbol: C9orf85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182505.3 | NP_872311.2 | 3 | 466 | ||
| XM_011518226.2 | XP_011516528.1 | 4 | 764 | |||
| GenBank mRNA | BC010179.2 | - | 2 | 355 | ||
| BC052375.1 | - | 3 | 305 | 6147448 | ||
| BQ431408.1 | - | 3 | 267 | |||

C9orf85
Genomic Map