siRNA Details
| Gene Symbol: C9orf135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010940.2 | NP_001010940.1 | 1, 2 | 234 | ||
| NM_001308084.1 | NP_001295013.1 | 1, 2 | 234 | |||
| XM_011518229.2 | XP_011516531.1 | 1, 2 | 235 | |||
| XM_011518230.2 | XP_011516532.1 | 1, 2 | 235 | |||
| XM_011518231.2 | XP_011516533.1 | 1, 2 | 235 | |||
| GenBank mRNA | BC136709.1 | - | 1, 2 | 211 | 9020699 | |
| BC150564.1 | - | 1, 2 | 212 | 40127624 | ||
| CD684431.1 | - | 1, 2 | 262 | |||

C9orf135
Genomic Map