siRNA Details
| Gene Symbol: FAM69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152421.3 | NP_689634.2 | 2 | 220 | ||
| XM_006716954.3 | XP_006717017.1 | 2 | 225 | |||
| XM_011518238.2 | XP_011516540.1 | 3 | 351 | |||
| XM_017014273.1 | XP_016869762.1 | 1 | 16 | |||
| GenBank mRNA | AJ420457.1 | - | 1 | 60 | ||
| BC032097.2 | - | 2 | 214 | 3161575 | ||
| BG700129.1 | - | 2 | 225 | |||

FAM69B
Genomic Map