siRNA Details
Gene Symbol: TEX13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195272.1 | NP_001182201.1 | 1 | 412 | ||
XM_005262352.3 | XP_005262409.1 | 1 | 661 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TEX13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195272.1 | NP_001182201.1 | 1 | 412 | ||
XM_005262352.3 | XP_005262409.1 | 1 | 661 |