siRNA Details
| Gene Symbol: VENTXP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001559.2 | 1 | 823 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

VENTXP1| Gene Symbol: VENTXP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001559.2 | 1 | 823 | |||