siRNA Details
| Gene Symbol: RBMXL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145346.1 | NP_001138818.1 | 1 | 2939 | ||
| GenBank mRNA | AK097568.1 | - | 1 | 1962 | ||
| AK301904.1 | - | 1 | 2939 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBMXL3| Gene Symbol: RBMXL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145346.1 | NP_001138818.1 | 1 | 2939 | ||
| GenBank mRNA | AK097568.1 | - | 1 | 1962 | ||
| AK301904.1 | - | 1 | 2939 | |||