siRNA Details
| Gene Symbol: ASB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202403.1 | NP_001189332.1 | 5 | 693 | ||
| NM_017873.3 | NP_060343.1 | 6 | 780 | |||
| NM_177999.2 | NP_821066.1 | 5 | 671 | |||
| GenBank mRNA | AK000555.1 | - | 6 | 777 | ||
| AK023004.1 | - | 6 | 726 | |||
| AK225618.1 | - | 5 | 668 | |||
| AK299193.1 | - | 5 | 575 | |||
| BC001719.1 | - | 5 | 626 | |||
| BC065913.1 | - | 6 | 683 | 5211288 | ||
| BX648923.1 | - | 5 | 1301 | |||

ASB6
Genomic Map