siRNA Details
| Gene Symbol: GATA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080473.4 | NP_536721.1 | 5 | 932 | ||
| XM_006723699.3 | XP_006723762.1 | 5 | 952 | |||
| GenBank mRNA | BC047790.1 | - | 5 | 932 | ||
| BC117356.1 | - | 4 | 895 | 40125907 | ||
| BC117358.1 | - | 4 | 895 | 40125909 | ||

GATA5
Genomic Map