siRNA Details
| Gene Symbol: SOGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080627.3 | NP_542194.2 | 13 | 3779 | ||
| NM_199181.2 | NP_954650.2 | 13 | 3052 | |||
| GenBank mRNA | AK126630.1 | - | 13 | 3052 | ||
| AK131491.1 | - | 9 | 2279 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOGA1| Gene Symbol: SOGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080627.3 | NP_542194.2 | 13 | 3779 | ||
| NM_199181.2 | NP_954650.2 | 13 | 3052 | |||
| GenBank mRNA | AK126630.1 | - | 13 | 3052 | ||
| AK131491.1 | - | 9 | 2279 | |||