siRNA Details
| Gene Symbol: TLDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080628.2 | NP_542195.1 | 4 | 449 | ||
| XM_017027672.1 | XP_016883161.1 | 4 | 439 | |||
| XM_017027674.1 | XP_016883163.1 | 3 | 289 | |||
| GenBank mRNA | AK123544.1 | - | 4 | 403 | ||
| BC127688.1 | - | 4 | 449 | 40127440 | ||
| BC130646.1 | - | 4 | 438 | 40146677 | ||
| BC130648.1 | - | 4 | 438 | 40146679 | ||

TLDC2
Genomic Map