siRNA Details
| Gene Symbol: SRXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080725.2 | NP_542763.1 | 2 | 2609 | ||
| GenBank mRNA | AF075053.1 | - | 1 | 454 | ||
| AK098418.1 | - | 2 | 1504 | |||
| AK127435.1 | - | 2 | 2454 | |||
| AL833944.1 | - | 2 | 2485 | |||
| AY427953.1 | - | 2 | 1504 | |||
| BC017001.1 | - | 2 | 2404 | |||
| BC032604.1 | - | 2 | 2440 | 5535883 | ||
| BC047707.1 | - | 2 | 2429 | 5735551 | ||

SRXN1
Genomic Map