siRNA Details
| Gene Symbol: SRXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080725.2 | NP_542763.1 | 2 | 573 | ||
| GenBank mRNA | AK127435.1 | - | 2 | 418 | ||
| AK312054.1 | - | 2 | 403 | |||
| AL833944.1 | - | 2 | 449 | |||
| BC017001.1 | - | 2 | 368 | |||
| BC032604.1 | - | 2 | 404 | 5535883 | ||
| BC047707.1 | - | 2 | 393 | 5735551 | ||
| BU170568.1 | - | 538 | ||||

SRXN1
Genomic Map