siRNA Details
| Gene Symbol: ROMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080748.2 | NP_542786.1 | 3 | 372 | ||
| XM_017027678.1 | XP_016883167.1 | 3 | 367 | |||
| GenBank mRNA | AK000548.1 | - | 3 | 277 | ||
| AM397244.1 | - | 3 | 271 | |||
| AM397245.1 | - | 3 | 277 | |||
| AM397246.1 | - | 2 | 352 | |||
| BC008488.1 | - | 388 | 4133274 | |||
| BC105281.1 | - | 3 | 281 | 6739248 | ||
| BM353595.1 | - | 1 | 84 | |||
| GU727625.1 | - | 3 | 281 | |||

ROMO1
Genomic Map