siRNA Details
| Gene Symbol: WFDC10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080753.2 | NP_542791.1 | 2 | 280 | ||
| XM_017027679.1 | XP_016883168.1 | 2 | 194 | |||
| GenBank mRNA | AY038182.1 | - | 2 | 280 | ||
| BC131579.1 | - | 2 | 246 | 40112102 | ||

WFDC10A
Genomic Map