siRNA Details
| Gene Symbol: LINC00934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024246.2 | 3, 4 | 1433 | |||
| GenBank mRNA | AK057497.1 | - | 3, 4 | 1433 | ||
| BC118632.1 | - | 1, 2 | 350 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00934| Gene Symbol: LINC00934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024246.2 | 3, 4 | 1433 | |||
| GenBank mRNA | AK057497.1 | - | 3, 4 | 1433 | ||
| BC118632.1 | - | 1, 2 | 350 | |||