siRNA Details
| Gene Symbol: LINC00459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX113955.1 | - | 2 | 104 | ||
| R46019.1 | - | 2 | 105 | |||
| T24087.1 | - | 2 | 93 | |||
| Z39313.1 | - | 2 | 90 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00459| Gene Symbol: LINC00459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX113955.1 | - | 2 | 104 | ||
| R46019.1 | - | 2 | 105 | |||
| T24087.1 | - | 2 | 93 | |||
| Z39313.1 | - | 2 | 90 | |||