siRNA Details
| Gene Symbol: LINC00355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057471.1 | - | 3 | 1014 | ||
| BC128161.1 | - | 2 | 247 | |||
| BC128162.1 | - | 2 | 244 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00355| Gene Symbol: LINC00355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057471.1 | - | 3 | 1014 | ||
| BC128161.1 | - | 2 | 247 | |||
| BC128162.1 | - | 2 | 244 | |||