siRNA Details
| Gene Symbol: LINC00343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046391.2 | 2 | 428 | |||
| NR_120418.1 | 2 | 428 | ||||
| GenBank mRNA | AK058053.1 | - | 2 | 389 | ||
| BC126908.1 | - | 2 | 367 | |||
| BC127249.1 | - | 2 | 367 | |||
| CD244335.1 | - | 2 | 407 | |||
| HY035387.1 | - | 2 | 429 | |||

LINC00343
Genomic Map