siRNA Details
| Gene Symbol: LINC00431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126378.1 | 2 | 545 | |||
| GenBank mRNA | AW086183.1 | - | 1 | 181 | ||
| BU630435.1 | - | 1 | 181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00431| Gene Symbol: LINC00431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126378.1 | 2 | 545 | |||
| GenBank mRNA | AW086183.1 | - | 1 | 181 | ||
| BU630435.1 | - | 1 | 181 | |||