siRNA Details
| Gene Symbol: LOC105370802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135680.1 | 4 | 463 | |||
| GenBank mRNA | BF725349.1 | - | 4 | 463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105370802| Gene Symbol: LOC105370802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135680.1 | 4 | 463 | |||
| GenBank mRNA | BF725349.1 | - | 4 | 463 | ||