siRNA Details
Gene Symbol: C15orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198784.1 | NP_001185713.1 | 2 | 517 | ||
GenBank mRNA | AK130862.1 | - | 2 | 494 | ||
BX648577.1 | - | 2 | 372 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C15orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198784.1 | NP_001185713.1 | 2 | 517 | ||
GenBank mRNA | AK130862.1 | - | 2 | 494 | ||
BX648577.1 | - | 2 | 372 |