siRNA Details
| Gene Symbol: DRAIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026979.1 | 3 | 497 | |||
| GenBank mRNA | BC029061.1 | - | 3 | 497 | ||
| BC104434.1 | - | 5 | 630 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DRAIC| Gene Symbol: DRAIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026979.1 | 3 | 497 | |||
| GenBank mRNA | BC029061.1 | - | 3 | 497 | ||
| BC104434.1 | - | 5 | 630 | |||