siRNA Details
| Gene Symbol: LOC145845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024264.1 | 5 | 662 | |||
| GenBank mRNA | AK023254.1 | - | 3 | 362 | ||
| BC133673.1 | - | 2 | 146 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC145845| Gene Symbol: LOC145845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024264.1 | 5 | 662 | |||
| GenBank mRNA | AK023254.1 | - | 3 | 362 | ||
| BC133673.1 | - | 2 | 146 | |||