siRNA Details
| Gene Symbol: VWA3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173615.4 | NP_775886.3 | 22, 23 | 2392 | ||
| XM_011545742.2 | XP_011544044.1 | 25, 26 | 2602 | |||
| XM_011545744.2 | XP_011544046.1 | 10, 11 | 1202 | |||
| XM_011545745.1 | XP_011544047.1 | 8, 9 | 970 | |||
| XM_011545746.2 | XP_011544048.1 | 8, 9 | 1073 | |||
| XM_017022947.1 | XP_016878436.1 | 26, 27 | 2675 | |||
| XM_017022948.1 | XP_016878437.1 | 9, 10 | 1774 | |||
| XM_017022949.1 | XP_016878438.1 | 7, 8 | 2045 | |||
| GenBank mRNA | AK128606.1 | - | 11, 12 | 1260 | ||
| BC109294.1 | - | 10, 11 | 1120 | |||
| BC109295.1 | - | 10, 11 | 1120 | |||

VWA3A
Genomic Map