siRNA Details
| Gene Symbol: CARMIL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013838.1 | NP_001013860.1 | 25 | 2616 | ||
| NM_001317026.1 | NP_001303955.1 | 26 | 2508 | |||
| XM_011522874.1 | XP_011521176.1 | 26 | 2646 | |||
| XM_011522875.2 | XP_011521177.2 | 26 | 2646 | |||
| XM_017022953.1 | XP_016878442.1 | 26 | 2646 | |||
| GenBank mRNA | AB113647.1 | - | 25 | 2616 | ||
| AK090421.1 | - | 3 | 3654 | |||
| AL832446.1 | - | 24 | 2447 | |||
| FJ026014.1 | - | 26 | 2388 | |||

CARMIL2
Genomic Map