siRNA Details
| Gene Symbol: ZNF534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143938.1 | NP_001137410.1 | 4 | 364 | ||
| NM_001143939.1 | NP_001137411.1 | 4 | 403 | |||
| GenBank mRNA | AB086839.1 | - | 4 | 364 | ||
| AB086840.1 | - | 4 | 403 | |||

ZNF534
Genomic Map