siRNA Details
| Gene Symbol: ZNF578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099694.1 | NP_001093164.1 | 6 | 1970 | ||
| XM_017026302.1 | XP_016881791.1 | 6 | 1990 | |||
| GenBank mRNA | AK055946.1 | - | 1 | 1148 | ||
| AK095562.1 | - | 1 | 665 | |||
| AK299106.1 | - | 6 | 1970 | |||
| BC103904.2 | - | 1 | 1138 | |||
| BC115704.1 | - | 1 | 1138 | |||

ZNF578
Genomic Map