siRNA Details
| Gene Symbol: TMEM190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139172.1 | NP_631911.1 | 2, 3 | 116 | ||
| GenBank mRNA | AF442729.1 | - | 2, 3 | 116 | ||
| BC128189.1 | - | 2, 3 | 116 | 40113868 | ||
| BC128190.1 | - | 2, 3 | 116 | 40113870 | ||

TMEM190
Genomic Map