siRNA Details
| Gene Symbol: ZNF524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153219.3 | NP_694951.1 | 2 | 359 | ||
| XM_011526487.2 | XP_011524789.1 | 2 | 600 | |||
| GenBank mRNA | BC007396.2 | - | 1 | 242 | ||
| BC014666.1 | - | 1 | 302 | 4864278 | ||
| BC067748.1 | - | 2 | 330 | 5313025 | ||
| BI756946.1 | - | 2 | 343 | |||
| BM790568.1 | - | 2 | 334 | |||
| HY097571.1 | - | 2 | 359 | |||

ZNF524
Genomic Map