siRNA Details
| Gene Symbol: CCDC155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144688.4 | NP_653289.3 | 4 | 506 | ||
| XM_005258549.3 | XP_005258606.1 | 4 | 506 | |||
| XM_011526488.2 | XP_011524790.1 | 4 | 506 | |||
| XM_011526489.2 | XP_011524791.1 | 4 | 2406 | |||
| XM_011526490.2 | XP_011524792.1 | 4 | 506 | |||
| XM_011526494.2 | XP_011524796.1 | 5 | 479 | |||
| XM_011526495.2 | XP_011524797.1 | 5 | 515 | |||
| XM_011526496.2 | XP_011524798.1 | 3 | 367 | |||
| XM_011526497.2 | XP_011524799.1 | 4 | 465 | |||
| XM_017026333.1 | XP_016881822.1 | 3 | 639 | |||
| GenBank mRNA | AK057220.1 | - | 4 | 475 | ||
| AK097696.1 | - | 5 | 1219 | |||
| BC029811.1 | - | 4 | 425 | 5171712 | ||
| DB024870.1 | - | 4 | 506 | |||

CCDC155
Genomic Map