siRNA Details
| Gene Symbol: LINC01533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110673.1 | 2 | 388 | |||
| GenBank mRNA | AA824408.1 | - | 2 | 218 | ||
| AK097493.1 | - | 2 | 388 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01533| Gene Symbol: LINC01533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110673.1 | 2 | 388 | |||
| GenBank mRNA | AA824408.1 | - | 2 | 218 | ||
| AK097493.1 | - | 2 | 388 | |||