siRNA Details
| Gene Symbol: FAM98C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174905.3 | NP_777565.3 | 4, 5 | 563 | ||
| GenBank mRNA | BC034383.1 | - | 1, 2 | 132 | ||
| BC117258.1 | - | 4, 5 | 563 | 40125809 | ||
| BC143830.1 | - | 4, 5 | 563 | 9052345 | ||
| BI871449.1 | - | 1, 2 | 95 | |||
| BM561971.1 | - | 4, 5 | 580 | |||

FAM98C
Genomic Map