siRNA Details
Gene Symbol: FAM98C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174905.3 | NP_777565.3 | 8 | 1055 | ||
GenBank mRNA | AK128745.1 | - | 5 | 3921 | ||
BC034383.1 | - | 6 | 593 | |||
BC080606.1 | - | 6 | 753 | |||
BC117258.1 | - | 8 | 1055 | 40125809 | ||
BC143830.1 | - | 8 | 1055 | 9052345 | ||
BX641942.1 | - | 1 | 234 |

More Information
Panther Classification:
Molecular Function -




