siRNA Details
| Gene Symbol: LINC00906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027318.2 | 2 | 2195 | |||
| GenBank mRNA | BC039341.1 | - | 2 | 2195 | ||
| BX647231.1 | - | 2 | 2193 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00906| Gene Symbol: LINC00906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027318.2 | 2 | 2195 | |||
| GenBank mRNA | BC039341.1 | - | 2 | 2195 | ||
| BX647231.1 | - | 2 | 2193 | |||