siRNA Details
| Gene Symbol: LINC00906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027318.2 | 2 | 331 | |||
| GenBank mRNA | BC039341.1 | - | 2 | 331 | ||
| BC101426.1 | - | 2 | 302 | |||
| BC101427.1 | - | 2 | 299 | |||
| BC101428.1 | - | 2 | 302 | |||
| BC122525.1 | - | 2 | 300 | |||
| BX647231.1 | - | 2 | 329 | |||

LINC00906
Genomic Map