siRNA Details
| Gene Symbol: ZNF681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138286.2 | NP_612143.2 | 4 | 627 | ||
| GenBank mRNA | AK056088.1 | - | 3 | 425 | ||
| AK122869.1 | - | 4 | 631 | |||
| BC062216.1 | - | 3 | 467 | |||
| BC136346.1 | - | 4 | 506 | 9020333 | ||

ZNF681
Genomic Map