siRNA Details
| Gene Symbol: LINC00905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024335.2 | 2 | 446 | |||
| NR_110321.1 | 2 | 156 | ||||
| GenBank mRNA | AK058057.1 | - | 2 | 405 | ||
| BC031284.1 | - | 2 | 156 | |||
| BC069223.1 | - | 2 | 378 | |||
| BC104221.2 | - | 1 | 343 | |||
| DB461577.1 | - | 2 | 446 | |||

LINC00905
Genomic Map