siRNA Details
| Gene Symbol: SAMD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010971.2 | NP_001010971.1 | 4 | 426 | ||
| NM_001134663.1 | NP_001128135.1 | 4 | 420 | |||
| NM_001134664.1 | NP_001128136.1 | 4 | 264 | |||
| XM_017000377.1 | XP_016855866.1 | 4 | 1094 | |||
| GenBank mRNA | AK056937.1 | - | 4 | 426 | ||
| AY604569.1 | - | 2 | 623 | |||
| BC037973.1 | - | 4 | 264 | |||
| DB459148.2 | - | 4 | 421 | |||

SAMD13
Genomic Map