siRNA Details
| Gene Symbol: SAMD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010971.2 | NP_001010971.1 | 4 | 529 | ||
| NM_001134663.1 | NP_001128135.1 | 4 | 523 | |||
| NM_001134664.1 | NP_001128136.1 | 4 | 367 | |||
| XM_017000377.1 | XP_016855866.1 | 4 | 1197 | |||
| GenBank mRNA | AK056937.1 | - | 4 | 529 | ||
| AY604569.1 | - | 2 | 726 | |||
| AY604570.1 | - | 2 | 423 | |||
| BC037973.1 | - | 4 | 367 | |||
| DB459148.2 | - | 4 | 524 | |||

SAMD13
Genomic Map