siRNA Details
| Gene Symbol: METTL11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136107.1 | NP_001129579.1 | 1, 2 | 221 | ||
| GenBank mRNA | BC157860.1 | - | 1, 2 | 221 | 9057079 | |
| BC171858.1 | - | 1, 2 | 221 | 9054512 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

METTL11B| Gene Symbol: METTL11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136107.1 | NP_001129579.1 | 1, 2 | 221 | ||
| GenBank mRNA | BC157860.1 | - | 1, 2 | 221 | 9057079 | |
| BC171858.1 | - | 1, 2 | 221 | 9054512 | ||